Learn More
Electron Microscopy Sciences K-kits Liquid TEM Kit, 2.0 µm Gap
SDP
Catalog No. 501928039
Encompass_Preferred
An Innovative Enclosed Specimen Holder for Liquid TEM. K-kits are sample holders designed to facilitate convenient TEM observation of liquid samples, allowing nanoobjects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized. With vacuum compatible sealing of liquids in electron-transmitting thickness, K-kits are micro reaction chambers for countless experiments in materials, chemical, and biological research.
Gap: 2.0 µm
Catalog No. 50-192-8039 Supplier Electron Microscopy Sciences Supplier No. K7260420
An Innovative Enclosed Specimen Holder for Liquid TEM. K-kits are sample holders designed to facilitate convenient TEM observation of liquid samples, allowing nanoobjects, aggregates, and agglomerates (NOAAs) in liquid samples to be characterized. With vacuum compatible sealing of liquids in electron-transmitting thickness, K-kits are micro reaction chambers for countless experiments in materials, chemical, and biological research.
Gap: 2.0 µm