Learn More
Hach Company Radiometer Analytical AL100 Replacement Salt-Bridge Double Liquid Junction (length = 70 mm, ceramic junction)
Catalog No. 501997541 Shop All Hach Company Products
Encompass
The Radiometer AL100 salt-bridge junction is a double-junction replacement salt bridge for 7.5 or 8 mm diameter reference electrodes. The AL100 has a total length of 70 mm. During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.
Catalog No. 50-199-7541 Supplier Hach Company Supplier No. B40A520
The Radiometer AL100 salt-bridge junction is a double-junction replacement salt bridge for 7.5 or 8 mm diameter reference electrodes. The AL100 has a total length of 70 mm. During measurement, the salt bridge junction (also called the liquid junction protection tube) prevents the reference electrode from being contaminated by the media and vice-versa. The salt-bridge junction consists of a glass tube capped by a ceramic liquid junction.