Learn More
THORLABS INC Liquid Light Guide, 03 mm Core 340 - 800 nm 4' 1.2 m Length
Catalog No. NC1597146
Encompass
Liquid Light Guide, 03 mm Core 340 - 800 nm 4' 1.2 m Length
Catalog No. NC1597146 Supplier THORLABS INC Supplier No. LLG034H
Liquid Light Guide, 03 mm Core 340 - 800 nm 4' 1.2 m Length