Learn More
MSC Nilfisk Mercury Vacuum, Electric, 12 Gallon Capacity, 4-Stage Filtration
Catalog No. 502838828
Encompass
This vacuum is designed to safely collect and retain liquid mercury spills and granular compounds, eliminating toxic vapors. Its four stages of filtration meet OSHA standards for inorganic mercury.
  • Collects mercury spills and granular compounds, eliminating toxic vapors.
  • Four-stage filtration meets OSHA standards for inorganic mercury.
  • Smooth-lined PVC hose reduces mercury droplet residue.
  • Centrifugal droplet separator collects liquid mercury.
  • Internal HEPA filter captures 99.97% of particles down to 0.3 microns.
  • Activated carbon adsorbent filter for mercury vapors.
  • Microstatic diffuser protects motor from superfine carbon particles.
  • Detachable trolley for easy decontamination.
  • Positive twist safety latches prevent unauthorized disassembly.
Catalog No. 50-283-8828 Supplier MSC Supplier No. 35210400
Add to Cart
Add to Cart
Request a Quote
This vacuum is designed to safely collect and retain liquid mercury spills and granular compounds, eliminating toxic vapors. Its four stages of filtration meet OSHA standards for inorganic mercury.
  • Collects mercury spills and granular compounds, eliminating toxic vapors.
  • Four-stage filtration meets OSHA standards for inorganic mercury.
  • Smooth-lined PVC hose reduces mercury droplet residue.
  • Centrifugal droplet separator collects liquid mercury.
  • Internal HEPA filter captures 99.97% of particles down to 0.3 microns.
  • Activated carbon adsorbent filter for mercury vapors.
  • Microstatic diffuser protects motor from superfine carbon particles.
  • Detachable trolley for easy decontamination.
  • Positive twist safety latches prevent unauthorized disassembly.