Learn More
Agilent Technologies Liquid N2 Cryogenic Valve, 1 ea
Catalog No. NC2748111
Encompass
This liquid N2 cryogenic valve is specifically designed for use with series 7890 gas chromatography systems. It enables precise flow control and provides reliable performance under demanding cryogenic conditions.
  • Designed for use with liquid nitrogen
  • Ensures stable flow control and durability
  • Offers reliable performance in cryogenic environments
  • Maintains compatibility with series 7890 gas chromatography systems
Catalog No. NC2748111 Supplier Agilent Technologies Supplier No. G343060018
This liquid N2 cryogenic valve is specifically designed for use with series 7890 gas chromatography systems. It enables precise flow control and provides reliable performance under demanding cryogenic conditions.
  • Designed for use with liquid nitrogen
  • Ensures stable flow control and durability
  • Offers reliable performance in cryogenic environments
  • Maintains compatibility with series 7890 gas chromatography systems