Learn More
Mitegen IceOff Liquid Nitrogen (N2) Drizzler for Crystal Samples
SDP
Catalog No. 501938884
Encompass
The Mitegen IceOff Liquid Nitrogen Drizzler is a tool designed to remove ice from frozen crystal samples. It operates by applying a small, targeted stream of liquid nitrogen to the sample. This is ideal for cryo-crystallography applications requiring a clear, frost-free view of the crystal.
Catalog No. 50-193-8884 Supplier Mitegen Supplier No. APIICEOFF01
The Mitegen IceOff Liquid Nitrogen Drizzler is a tool designed to remove ice from frozen crystal samples. It operates by applying a small, targeted stream of liquid nitrogen to the sample. This is ideal for cryo-crystallography applications requiring a clear, frost-free view of the crystal.