Learn More
Sigma Aldrich Fine Chemicals Biosciences Liquid nitrogen cooled min1EA
Catalog No. 502905888
Encompass
freeze and Process tiny tissue samples within a microcentrifuge tube. Stainless steel mortar has a welded fixture to hold a 1.5 mL microcentrifuge tube. The mortar is suspended above a stainless steel cavity which holds the liquid nitrogen and is nested in a heavily insulated blue polyethylene housing.Permits grinding of smaller amounts of tissue directly in a 1.5 mL microcentrifuge tube resulting in reduced sample loss and collection timePrevents tube from cracking by eliminating direct contact with liquid nitrogenIncludes 5 in. long stainless steel pestle with W x D 10.2 cm x 1.9 cm polyethylene handleHousing overall dimensions D x H 16.5 cm x 11.4 cm suction feet keep it stable on the workbenchRemovable insulated mortar handle 10.2 cm long
Catalog No. 50-290-5888 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7563771EA
freeze and Process tiny tissue samples within a microcentrifuge tube. Stainless steel mortar has a welded fixture to hold a 1.5 mL microcentrifuge tube. The mortar is suspended above a stainless steel cavity which holds the liquid nitrogen and is nested in a heavily insulated blue polyethylene housing.Permits grinding of smaller amounts of tissue directly in a 1.5 mL microcentrifuge tube resulting in reduced sample loss and collection timePrevents tube from cracking by eliminating direct contact with liquid nitrogenIncludes 5 in. long stainless steel pestle with W x D 10.2 cm x 1.9 cm polyethylene handleHousing overall dimensions D x H 16.5 cm x 11.4 cm suction feet keep it stable on the workbenchRemovable insulated mortar handle 10.2 cm long