Learn More
Taylor Wharton Cryogenic Storage; LD4, 4 L
Catalog No. NC1509729
Encompass_Preferred
State-of-the-art construction and advances insulation materials for high thermal efficiency. Ribbed, high-strength aluminum body, magniformed neck tube design, and durable paint. Easy to operate--light-weight, snap-on cap and precise-fitting neck tube assure tight closure and easy access. Large, convenient handles for easy maneuvering.
Catalog No. NC1509729 Supplier Taylor Wharton Supplier No. 4LDB
State-of-the-art construction and advances insulation materials for high thermal efficiency. Ribbed, high-strength aluminum body, magniformed neck tube design, and durable paint. Easy to operate--light-weight, snap-on cap and precise-fitting neck tube assure tight closure and easy access. Large, convenient handles for easy maneuvering.