Learn More
Thomas Scientific LIQUID NITROGEN STORAGE DEWAR, 21 LITER
SDP
Catalog No. 501458023
Encompass_Preferred
The storage and dispensing of liquid nitrogen can now be accomplished safely and efficiently with liquid dewars from American Biotech Supply. These super insulated dewars have minimal evaporation rates and are available for pouring, pressurized dispensing and dipping of liquid nitrogen. These high efficiency, super insulated dewars are the most convenient, economical way to store and dispense liquid nitrogen.
Catalog No. 50-145-8023 Supplier Thomas Scientific Supplier No. 1158P14
The storage and dispensing of liquid nitrogen can now be accomplished safely and efficiently with liquid dewars from American Biotech Supply. These super insulated dewars have minimal evaporation rates and are available for pouring, pressurized dispensing and dipping of liquid nitrogen. These high efficiency, super insulated dewars are the most convenient, economical way to store and dispense liquid nitrogen.