Learn More
Electron Microscopy Sciences Thermo-Flask Liquid Nitrogen Transfer Vessel, 4.5L, 6 I.D., 7.25 O.D., 13.75 height
SDP
Catalog No. 501899057
Encompass_Preferred
Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2, and thus have a broad range of applications in industrial, educational, and clinical fields. Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5 mm of Hg to provide maximum coolant retention. Corrosion-resistant stainless steel exterior. Carrying handle, vented lid secured by clamps.
Capacity: 4.5 L
I.D.: 6
O.D.: 7.25
Height: 13.75
Catalog No. 50-189-9057 Supplier Electron Microscopy Sciences Supplier No. 6189104
Thermo-Flask provides safe extended temperature retention for specimens stored in liquid nitrogen or solid CO2, and thus have a broad range of applications in industrial, educational, and clinical fields. Thermo-Flask features borosilicate glass inner liners which have been evacuate to 1x10-5 mm of Hg to provide maximum coolant retention. Corrosion-resistant stainless steel exterior. Carrying handle, vented lid secured by clamps.
Capacity: 4.5 L
I.D.: 6
O.D.: 7.25
Height: 13.75