Learn More
Cole-Parmer LIQUID NITROGEN WI-FI 2 PROBES
Supplier:  Cole-Parmer 1800109
Encompass_Preferred
Traceable Liquid Nitrogen Wi-Fi Data Logger Compatible with TraceableLIVE Cloud Service; 2 SS Probes
Catalog No. NC1970088