Learn More
Macherey-Nagel Liquid RNase A, 100 mg/mL Concentration, Ready-to-Use Liquid, 2.5 mL
Catalog No. NC2681701
Encompass_Preferred
MACHEREY-NAGEL Liquid RNase A is a ready-to-use enzyme solution designed for RNA digestion in molecular biology. This highly concentrated RNase A facilitates efficient sample preparation for cell lysis and nucleic acid digestion, eliminating the need for prior dissolution. Its compatibility with NucleoSpin™ and NucleoBond™ kits ensures streamlined workflows. With a concentration of 100 mg/mL and a shelf life of 24 months, it stands out for its quality and effectiveness.
Catalog No. NC2681701 Supplier Macherey-Nagel Supplier No. 740397
MACHEREY-NAGEL Liquid RNase A is a ready-to-use enzyme solution designed for RNA digestion in molecular biology. This highly concentrated RNase A facilitates efficient sample preparation for cell lysis and nucleic acid digestion, eliminating the need for prior dissolution. Its compatibility with NucleoSpin™ and NucleoBond™ kits ensures streamlined workflows. With a concentration of 100 mg/mL and a shelf life of 24 months, it stands out for its quality and effectiveness.