Learn More
Sigma Aldrich Fine Chemicals Biosciences Liquid scintillation vials
Catalog No. 502905605
Encompass
Reliable Wheaton 180 brand low-potassium glass. White 22Amm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.
Catalog No. 50-290-5605 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z1905191PAK
Reliable Wheaton 180 brand low-potassium glass. White 22Amm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.