Learn More
Sigma Aldrich Fine Chemicals Biosciences Liquid scintillation vials
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z1905431PAK
Encompass
Reliable Wheaton 180 brand low-potassium glass. White 22Amm caps have tops suitable for writing. Vials with caps attached are packed in individual cells in trays of 100.
Catalog No. 50-290-4005