Learn More
Meridian Life Science Liquid Stable Flex DNA qPCR, 40ML
SDP
Catalog No. 5000856382
Encompass_Preferred
Liquid Stable Flex DNA qPCR Mix is a combination of the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with an antibody-mediated hot start polymerase and containing dUTP. Liquid Stable Flex DNA qPCR Mix has been designed to be stable for at least 12 months at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Flex DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.
Catalog No. 50-008-56382 Supplier Meridian Life Science Supplier No. MDX36040
Liquid Stable Flex DNA qPCR Mix is a combination of the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with an antibody-mediated hot start polymerase and containing dUTP. Liquid Stable Flex DNA qPCR Mix has been designed to be stable for at least 12 months at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Flex DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.