Learn More
Meridian Life Science Liquid Stable Flex RNA/DNA, 4ML
SDP
Catalog No. 5000856455
Encompass_Preferred
Liquid Stable Flex RNA/DNA qPCR Mix is a combination of the latest advances in buffer chemistry containing dUTP and PCR enhancers and stabilizers, together with reverse transcriptase and an antibody-mediated hot start polymerase. Liquid Stable Flex RNA/DNA qPCR Mix has been designed to be stable at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Flex RNA/DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.
Catalog No. 50-008-56455 Supplier Meridian Life Science Supplier No. MDX9884
Liquid Stable Flex RNA/DNA qPCR Mix is a combination of the latest advances in buffer chemistry containing dUTP and PCR enhancers and stabilizers, together with reverse transcriptase and an antibody-mediated hot start polymerase. Liquid Stable Flex RNA/DNA qPCR Mix has been designed to be stable at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Flex RNA/DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.