Learn More
Meridian Life Science Liquid Stable Ready DNA qPCR, 4ML
SDP
Catalog No. 5000856454
Encompass_Preferred
Liquid Stable Ready DNA qPCR Mix is a combination of the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with an antibody-mediated hot start polymerase and containing dUTP. Liquid Stable Ready DNA qPCR Mix has been designed to enable pre-assembled qPCR assay (MDX364 + assay oligonucleotides) be stable for at least 12 months at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Ready DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.
Catalog No. 50-008-56454 Supplier Meridian Life Science Supplier No. MDX3644
Liquid Stable Ready DNA qPCR Mix is a combination of the latest advances in buffer chemistry and PCR enhancers and stabilizers, together with an antibody-mediated hot start polymerase and containing dUTP. Liquid Stable Ready DNA qPCR Mix has been designed to enable pre-assembled qPCR assay (MDX364 + assay oligonucleotides) be stable for at least 12 months at ambient temperature. In addition, it has been optimized for highly reproducible and accurate assay, delivering exceptional performance in multiplex assays, even under fast thermal cycling conditions. Liquid Stable Ready DNA qPCR Mix formulation is compatible to applications where reduction of false positives from cross-over contamination is critical.