Learn More
Surmodics Inc 450 nm Liquid Stop Solution for TMB Microwell Substrates, 1L
SDP
Catalog No. NC9141468
Encompass_Preferred
Liquid Stop Reagent for TMB Microwell, 1 x 1L, 450nm Changes Blue TMB Reaction to Yellow to Read at 450nm, in 20 to 60 minutes
Catalog No. NC9141468 Supplier Surmodics Inc Supplier No. LSTP100001
Liquid Stop Reagent for TMB Microwell, 1 x 1L, 450nm Changes Blue TMB Reaction to Yellow to Read at 450nm, in 20 to 60 minutes