Learn More
MilliporeSigma Steridilutor NEO Device
Supplier:  MilliporeSigma TZA000010
Encompass
The Steridilutor NEO device for liquid transfer kit enables to dilute liquids from open ampoules into a diluent container with septum. For liquid antibiotics, a dilution step is strongly recommended to reduce the concentration of the antimicrobial agent. This will reduce the possibility of binding of the agent to the filter.
Catalog No. NC1141077