Learn More
Valiteq LIQUID TSB-2X 30ML 12/PK
SDP
Catalog No. NC9891899
Encompass_Preferred
30 Ml Vials Of Liquid Tsb - 2X. 12 Vials Per Package. The Vm-30 Contains 30Ml Of Double-Strength Trypticase-Soy Broth To Facilitate The Use Of The Product In More Complex Validations Requiring Further Dilution And Transfer.
Catalog No. NC9891899 Supplier Valiteq Supplier No. VM 30
30 Ml Vials Of Liquid Tsb - 2X. 12 Vials Per Package. The Vm-30 Contains 30Ml Of Double-Strength Trypticase-Soy Broth To Facilitate The Use Of The Product In More Complex Validations Requiring Further Dilution And Transfer.