Learn More
Cliawaived Inc Liquid QC DipStrip Urinalysis Controls
SDP
Catalog No. NC1859815
Encompass_Preferred
Bi-Level (3 x 2 x 15mL)Liquid urinalysis control for use with all Siemens Urinalysis Multistix products, Roche Chemstrip, CLINITEK Microalbumin, Multistix PRO 10LS, Siemens Acetest, ClinTest and Bayer Ictotest.Refrigerated item for product stability.
This item is not returnable. View return policy
Catalog No. NC1859815 Supplier Cliawaived Inc Supplier No. CQ97201
Bi-Level (3 x 2 x 15mL)Liquid urinalysis control for use with all Siemens Urinalysis Multistix products, Roche Chemstrip, CLINITEK Microalbumin, Multistix PRO 10LS, Siemens Acetest, ClinTest and Bayer Ictotest.Refrigerated item for product stability.