Learn More
Electron Microscopy Sciences Paper Liquid Wicks, 28 mm Size Fine
SDP
Catalog No. 509053392
Encompass
Theses paper wicks offer a great deal of absorbency with small tips that foam-tipped or cotton-tipped applicators can not substitute. Each box contains 300, 28 mm long high quality, tapered paper wicks. Size Fine (F) wicks are ideal for cleaning MiroMount. The thick end of these wicks fit into standard 0.7mm mechanical pencil.
Catalog No. 50-905-3392 Supplier Electron Microscopy Sciences Supplier No. 60160F
Theses paper wicks offer a great deal of absorbency with small tips that foam-tipped or cotton-tipped applicators can not substitute. Each box contains 300, 28 mm long high quality, tapered paper wicks. Size Fine (F) wicks are ideal for cleaning MiroMount. The thick end of these wicks fit into standard 0.7mm mechanical pencil.