Learn More
Cole-Parmer Alconox Liquinox 1201 Critical Cleaning Liquid Detergent; 4 x 1 gal. Bottles/Cs
Catalog No. NC9480666 Shop All Cole-Parmer Products
Encompass_Preferred
White powder Alconox, Alcojet, and Tergazyme cleaners rinses off completelywith no residue. Alconox cleaner is biodegradable; Alcojet cleaner is low-sudsing; Tergazyme cleaner combines the original Alconox power with protein enzymes. Liquinox cleaner is biodegradableuse to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA compliant.
Catalog No. NC9480666 Supplier Cole-Parmer Supplier No. 1777540
White powder Alconox, Alcojet, and Tergazyme cleaners rinses off completelywith no residue. Alconox cleaner is biodegradable; Alcojet cleaner is low-sudsing; Tergazyme cleaner combines the original Alconox power with protein enzymes. Liquinox cleaner is biodegradableuse to remove soil, grease, blood, chemicals, and solvents. All cleaner are safe, odorless, and FDA compliant.