Learn More
Perkin Elmer US LLC Liquids Retainer with Volatiles Cover
Supplier:  Perkin Elmer US LLC L1272321
Encompass_Preferred
The Liquids Retainer plate is used on Pike ATR accessories that are compatible with the Spectrum One/100/400/65, Spectrum Two, and Spectrum 3/Frontier spectrometers.
Catalog No. 50-211-3233