Learn More
Perkin Elmer US LLC Liquids Sampling Starter Pack
Catalog No. 502113616
Encompass_Preferred
This starter pack equips you with everything you need to prepare liquids and mulls for FT-IR analysis.
Catalog No. 50-211-3616 Supplier Perkin Elmer US LLC Supplier No. L1272268
This starter pack equips you with everything you need to prepare liquids and mulls for FT-IR analysis.