Learn More
Research Products International Corp Liqui-Nox Liquid Detergent, 1 Gallon
SDP
Catalog No. 50476436
Encompass
A biodegradable phosphate free liquid detergent with high cleaning power. Liqui-Nox is a special formulation of anionic and non-ionic detergents and wetting agents. Works quickly and effectively in hard, soft, cold or hot water. Spotlessly cleans all glass, metal, plastic, rubber and hard surface materials. Leaves no residue.
Catalog No. 50-476-436 Supplier Research Products International Corp Supplier No. 114011
A biodegradable phosphate free liquid detergent with high cleaning power. Liqui-Nox is a special formulation of anionic and non-ionic detergents and wetting agents. Works quickly and effectively in hard, soft, cold or hot water. Spotlessly cleans all glass, metal, plastic, rubber and hard surface materials. Leaves no residue.