Learn More
Sigma Aldrich Fine Chemicals Biosciences LiquinoxR phosphate-free l
Supplier:  Sigma Aldrich Fine Chemicals Biosciences Z7429163.8L
Encompass
Concentrated anionic detergent for manual and ultrasonic cleaning. free rinsing to give you reliable results without interfering residues. Extremely mild and completely soluble in hard and soft water. USDA authorized. Dilute 1100. pH 8.5
Catalog No. 50-290-0571