Learn More
Cayman Chemical (±)-Liquiritigenin, Purity: ≥98%, CAS: 69097-97-8, 100 mg
SDP
Catalog No. 502939829
Encompass_Preferred
A flavonoid with anticancer activity; cytotoxic to HL-60, HCT8, and B16 cancer cell lines (IC50s = 22.5, 19.5, and 5 μg/ml, respectively)
Catalog No. 50-293-9829 Supplier Cayman Chemical Supplier No. 28823100 MG
A flavonoid with anticancer activity; cytotoxic to HL-60, HCT8, and B16 cancer cell lines (IC50s = 22.5, 19.5, and 5 μg/ml, respectively)