Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Liquiritigenin-7-O-β-D-glucopyranosyl-(1→2)-β-D-apiofuranoside | 135432-48-3 | 10 MG
SDP

Catalog No. 5000418361
Encompass_Preferred

Liquiritigenin-7-O-β-D-glucopyranosyl-(1→2)-β-D-apiofuranoside, also known as Liquiritigenin-7-apiosylglucoside, is a flavonoid isolated from the roots of Glycyrrhiza. It is recognized for exhibiting weaker cytotoxicity against various tumor cells and normal cells. This product is strictly for research use only and not intended for sale to patients.

  • Source: Isolated from the roots of Glycyrrhiza.
  • Activity: Flavonoid with weaker cytotoxicity against several tumor cells and normal cells.
  • Purity: 99.74% confirmed.
  • Appearance: Solid, light yellow to yellow in color.
  • In vitro solubility: 100 mg/mL in DMSO (181.65 mM).
  • In vivo solubility: Can be dissolved in various solvent systems such as 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline; 10% DMSO, 90% (20% SBE-β-CD in Saline); or 10% DMSO, 90% Corn Oil, with solubilities generally ≥ 2.5 mg/mL (4.54 mM).

Catalog No. 50-004-18361 Supplier Medchemexpress LLC Supplier No. HYN698610MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Liquiritigenin-7-O-β-D-glucopyranosyl-(1→2)-β-D-apiofuranoside, also known as Liquiritigenin-7-apiosylglucoside, is a flavonoid isolated from the roots of Glycyrrhiza. It is recognized for exhibiting weaker cytotoxicity against various tumor cells and normal cells. This product is strictly for research use only and not intended for sale to patients.

  • Source: Isolated from the roots of Glycyrrhiza.
  • Activity: Flavonoid with weaker cytotoxicity against several tumor cells and normal cells.
  • Purity: 99.74% confirmed.
  • Appearance: Solid, light yellow to yellow in color.
  • In vitro solubility: 100 mg/mL in DMSO (181.65 mM).
  • In vivo solubility: Can be dissolved in various solvent systems such as 10% DMSO, 40% PEG300, 5% Tween-80, 45% Saline; 10% DMSO, 90% (20% SBE-β-CD in Saline); or 10% DMSO, 90% Corn Oil, with solubilities generally ≥ 2.5 mg/mL (4.54 mM).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.