Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Liquiritigenin, Thermo Scientific™

Catalog No. AC469812500
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC469812500 250 mg
AC469810010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC469812500 Supplier Thermo Scientific Chemicals Supplier No. 469812500
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart
This item is not returnable. View return policy

Specifications

Alpha Vector LIQUIRITIGENIN
Chemical Name or Material Liquiritigenin
CAS 578-86-9
Color Beige to White
Boiling Point 530°C (760 mmHg)
Infrared Spectrum Conforms
Packaging Glass bottle
Molecular Formula C15 H12 O4
Quantity 250 mg
MDL Number 00287289
Synonym 4 ',7-Dihydroxyflavanone
Molecular Weight (g/mol) 256.26
Physical Form Crystals or Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.