Learn More
TARGETMOL CHEMICALS INC LIQUIRITIN 25MG
SDP
Catalog No. 502577030
Encompass_Preferred
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Liquiritin (Liquiritigenin-4'-O-glucoside) (LIQ) is a main component among the licorice flavonoids and possesses anti-inflammatory and anti-cancer abilities. purity: 98%
Catalog No. 50-257-7030 Supplier TARGETMOL CHEMICALS INC Supplier No. T289925MG
Also available in 5 mg 10 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Liquiritin (Liquiritigenin-4'-O-glucoside) (LIQ) is a main component among the licorice flavonoids and possesses anti-inflammatory and anti-cancer abilities. purity: 98%