Learn More
Medchemexpress LLC Liquiritin apioside | 74639-14-8 | MFCD09260034 | 100.0% | 550.51 g·mol⁻¹ | C26H30O13 | 10 MG
SDP
Catalog No. 5000296198
Encompass_Preferred
Liquiritin apioside is a flavonoid O-glycoside used as a research reagent and reported as an orally active inhibitor of the TRPV1 receptor. It selectively inhibits the laryngeal chemoreflex and is applied in studies of respiratory-related pharmacology and analytical research.
  • Orally active TRPV1 receptor inhibitor.
  • Suitable for respiratory-related pharmacology research.
  • High chemical purity and analytical-grade quality.
  • Available in multiple package sizes, including 10 mg.
  • Characterized by CAS 74639-14-8 and formula C26H30O13.
Catalog No. 50-002-96198 Supplier Medchemexpress LLC Supplier No. HYN147110MG
Liquiritin apioside is a flavonoid O-glycoside used as a research reagent and reported as an orally active inhibitor of the TRPV1 receptor. It selectively inhibits the laryngeal chemoreflex and is applied in studies of respiratory-related pharmacology and analytical research.
  • Orally active TRPV1 receptor inhibitor.
  • Suitable for respiratory-related pharmacology research.
  • High chemical purity and analytical-grade quality.
  • Available in multiple package sizes, including 10 mg.
  • Characterized by CAS 74639-14-8 and formula C26H30O13.