Learn More
BURKLE INC LiquiSampler, PP, 5.07 oz (150 ml) volume, 23.62 in (60 cm), Ø 1.26 in (32 mm)
Catalog No. 5000026037
Encompass_Preferred
LiquiSampler for liquids, according to DIN EN ISO 15528. Made of ultra pure and chemically inert PTFE/FEP or PP for contamination-free sampling.
Sampling from open and closed containers, barrels, tanks, silos, water courses. Suitable for all commercially available barrels and containers with opening of at least 1.26 in. (32 mm) (1.26 in.) in diameter.
Easy cleaning - all surfaces are free of pores and crevices, preventing accumulation of dirt. Only round screw threads proven in foodstuffs hygiene are used. Disassembly and cleaning are extremely simple.
Catalog No. 50-000-26037 Supplier BURKLE INC Supplier No. 53301060
LiquiSampler for liquids, according to DIN EN ISO 15528. Made of ultra pure and chemically inert PTFE/FEP or PP for contamination-free sampling.
Sampling from open and closed containers, barrels, tanks, silos, water courses. Suitable for all commercially available barrels and containers with opening of at least 1.26 in. (32 mm) (1.26 in.) in diameter.
Easy cleaning - all surfaces are free of pores and crevices, preventing accumulation of dirt. Only round screw threads proven in foodstuffs hygiene are used. Disassembly and cleaning are extremely simple.