Learn More
National Diagnostics LIQUISCINT 20 LT
SDP
Catalog No. 5089990170
Encompass
Liquiscint, Economical, ready-to-use liquid scintillation solution designed for aqueous and organic samples, 20 LT
Catalog No. 50-899-90170 Supplier National Diagnostics Supplier No. LS12120LTR
Liquiscint, Economical, ready-to-use liquid scintillation solution designed for aqueous and organic samples, 20 LT