Learn More
World Precision Instrument LiquiVet Rapid Tissue Adhesive
SDP
Catalog No. 502539340
Encompass
Veterinary use only. LiquiVet monomeric formulation forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Comes in 3 g dispensing bottles with five applicators.
Catalog No. 50-253-9340 Supplier World Precision Instrument Supplier No. 504561
Veterinary use only. LiquiVet monomeric formulation forms a thin, waterproof adhesion to bond tissue in a variety of vet procedures. Comes in 3 g dispensing bottles with five applicators.