Learn More
Medchemexpress LLC Liraglutide-13C5,15N tetraTFA | 204656-20-2 | 97.7% | 4214.26 | 5 MG
SDP
Catalog No. 5000426695
Encompass_Preferred
Liraglutide-13C5,15N tetraTFA is the 13C and 15N labeled Liraglutide. Liraglutide is a glucagon-like peptide-1 (GLP-1) receptor agonist used for the study of type 2 diabetes mellitus. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Aids in understanding pharmacokinetic and metabolic profiles of drugs.
Catalog No. 50-004-26695 Supplier Medchemexpress LLC Supplier No. HYP0014S15MG
Liraglutide-13C5,15N tetraTFA is the 13C and 15N labeled Liraglutide. Liraglutide is a glucagon-like peptide-1 (GLP-1) receptor agonist used for the study of type 2 diabetes mellitus. It is for research use only and not sold to patients.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Incorporates stable heavy isotopes for quantitation during drug development.
  • Aids in understanding pharmacokinetic and metabolic profiles of drugs.