Learn More
Medchemexpress LLC Liraglutide-13C5,15N tetraTFA | 97.7% | 4214.26 | 1 MG
SDP
Catalog No. 5000426213
Encompass_Preferred
Liraglutide-13C5,15N tetraTFA is the 13C and 15N labeled version of Liraglutide. Liraglutide is a glucagon-like peptide-1 (GLP-1) receptor agonist, which is used for the study of type 2 diabetes mellitus.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-26213 Supplier Medchemexpress LLC Supplier No. HYP0014S11MG
Liraglutide-13C5,15N tetraTFA is the 13C and 15N labeled version of Liraglutide. Liraglutide is a glucagon-like peptide-1 (GLP-1) receptor agonist, which is used for the study of type 2 diabetes mellitus.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.