Learn More
Carbosynth LLC. Liraglutide, 1mg, MW: (M.W. 3751.29), Molecular formula: C17H265N43O51, CAS# [204656-20-2]
SDP
Supplier:  Carbosynth LLC. PGL3781PI1MG
Encompass_Preferred
Synonyms: H-His-Ala-Glu-Gly-Thr-Phe-Thr-Ser-Asp-Val-Ser-Ser-Tyr-Leu-Glu-Gly-Gln-Ala-Ala-Lys(γ-Glu-palmitoyl)-Glu-Phe-Ile-Ala-Trp-Leu-Val-Arg-Gly-Arg-Gly-OH Lys(γ-Glu-palmitoyl)26,Arg34)-GLP-1 (7-37), Application: Glucagon-Like-Peptide-1 (GLP-1) Receptor Agonist, Storage Temperature: -20°C, References: J.J.Neumiller, J. Am. Pharm. Assoc., 49, S16 (2009), M.C.Rossi and A.Nicolucci, Acta Biomed., 80, 93 (2009), C.F.Deacon, Vasc. Health Risk Manag., 5, 199 (2009)
Catalog No. 50-168-6543