Learn More
Advanced Chemblocks Inc Liraglutide 1G, white to almost white powder, 98% purity, 3751 g/mol
Supplier: Advanced Chemblocks Inc 104371G
Liraglutide is a synthetic glucagon-like peptide-1 (GLP-1) receptor agonist designed for scientific research. It mimics human GLP-1 with modifications that enhance its stability and prolong its action. This product is primarily used for therapeutic applications in managing diabetes and obesity, offering significant benefits in glycemic control and weight management.
- Potent GLP-1 receptor agonist
- Improves glucose-dependent insulin secretion
- Suppresses postprandial glucagon secretion
- Slows gastric emptying
- Demonstrates efficacy in weight loss
- Half-life of approximately 13 hours
- Available as a white to almost white powder
- Soluble in DMSO and other solvents
- Store at -20 °C for long-term
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.