Learn More
Selleck Chemical LLC Liraglutide 1mg 204656-20-2 Victoza, Liraglutida, Liraglutidum, NN2211, NN-2211
SDP
Catalog No. 501365858
Encompass_Preferred
Liraglutide (Victoza, Liraglutida, Liraglutidum, NN2211) is a long-acting glucagon-like peptide-1(GLP-1) receptor agonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5858 Supplier Selleck Chemical LLC Supplier No. S82561MG
Liraglutide (Victoza, Liraglutida, Liraglutidum, NN2211) is a long-acting glucagon-like peptide-1(GLP-1) receptor agonist. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.