Learn More
AdipoGen Liraglutide (25 mg)
SDP
Supplier:  AdipoGen AGCP30034M025
Encompass_Preferred
Liraglutide (25 mg), Adipogen Life Sciences. Chemical. CAS: 204656-20-2. Formula: C172H265N43O51. MW: 3751.2. Synthetic. Long-acting acylated glucagon-like peptide-1 (GLP-1) receptor agonist. Antidiabetic and antiobesity agent used clinically to treat type 2 diabetes mellitus. Binding to GLP-1R, activates AMP-activated protein kinase, consequently stimulates insulin secretion in pancreatic beta cells and suppresses glucagon secretion in a glucose-dependent manner. Improves control of blood glucose and consequently modulates appetite and body weight. Inhibits beta cell apoptosis and improves beta cell mass. Shown to ameliorate glycometabolism and insulin resistance through the upregulation of GLUT4. Neuroprotective. Prevents neurodegenerative processes in pathologies such as Alzheimer's and Parkinsons Disease. Anti-inflammatory agent. Exerts cardioprotective roles via activating prosurvival pathways and suppressing inflammation.
Catalog No. 50-168-7409