Learn More
Eurofins Discoverx cAMP Hunter Liraglutide Bioassay Kit, 10-Plate. (95-0062Y2-00100)
Catalog No. 502002604
Encompass
The cAMP Hunter Liraglutide Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses Liraglutide based activation of GLP-1R GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GLP-1R, a naturally Gs coupled GPCR. Upon activation of GLP-1R, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.
Catalog No. 50-200-2604 Supplier Eurofins Discoverx Supplier No. 950062Y200100
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter Liraglutide Bioassay kit provides an easy-to-use cell based assay to measure drug potency and detect neutralizing antibodies. This bioassay assesses Liraglutide based activation of GLP-1R GPCR activity via detection of levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type GLP-1R, a naturally Gs coupled GPCR. Upon activation of GLP-1R, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. Bioassay kits are a convenient, ready-to-use format which contains all materials needed to run the assay, including single-use vials of cryopreserved cells, cell plating media, control agonist, detection reagent, and assay plates. For detailed product information, User manual and COA, visit www.discoverx.com/bioassay.