Learn More
Medchemexpress LLC Liraglutide TFA | 204656-20-2 | 99.6% | 3751.20 (free base) | 1 MG
SDP
Catalog No. 5000425051
Encompass_Preferred
Liraglutide (TFA) is an agonist of glucagon-like peptide 1 receptor (GLP-1). It can activate GLP-1, leading to insulin release in the presence of increased glucose concentration, and also reduces glucagon secretion in a glucose-dependent manner. It can be used in research on type 2 diabetes.
  • Activates GLP-1 receptor
  • Leads to insulin release
  • Reduces glucagon secretion
  • Suitable for type 2 diabetes research
Catalog No. 50-004-25051 Supplier Medchemexpress LLC Supplier No. HYP0014B1MG
Liraglutide (TFA) is an agonist of glucagon-like peptide 1 receptor (GLP-1). It can activate GLP-1, leading to insulin release in the presence of increased glucose concentration, and also reduces glucagon secretion in a glucose-dependent manner. It can be used in research on type 2 diabetes.
  • Activates GLP-1 receptor
  • Leads to insulin release
  • Reduces glucagon secretion
  • Suitable for type 2 diabetes research