Learn More
Cayman Chemical Liraglutide (acetate), Purity: ≥95%, CAS: 1997361-86-0, 10 mg
SDP
Catalog No. 502935738
Encompass_Preferred
A GLP-1R agonist; increases cAMP accumulation in BHK cells expressing GLP-1R (EC50 = 61 pM); inhibits cytokine- and free fatty acid-induced apoptosis of primary neonatal rat pancreatic β-cells in vitro by greater than 95 and 50%, respectively at 100-1,000 nM; decreases calorie intake, shifts food preference to a higher ratio of chow to candy, reverses weight and fat gains, and increases insulin sensitivity in a rat model of diet-induced obesity at 0.2 mg/kg, s.c., twice per day
Catalog No. 50-293-5738 Supplier Cayman Chemical Supplier No. 2472710 MG
A GLP-1R agonist; increases cAMP accumulation in BHK cells expressing GLP-1R (EC50 = 61 pM); inhibits cytokine- and free fatty acid-induced apoptosis of primary neonatal rat pancreatic β-cells in vitro by greater than 95 and 50%, respectively at 100-1,000 nM; decreases calorie intake, shifts food preference to a higher ratio of chow to candy, reverses weight and fat gains, and increases insulin sensitivity in a rat model of diet-induced obesity at 0.2 mg/kg, s.c., twice per day