Learn More
Selleck Chemical LLC Lirametostat CPI-1205-2mg
SDP
Catalog No. 502389922
Encompass_Preferred
Lirametostat (CPI-1205) is an orally available selective inhibitor of the histone lysine methyltransferase EZH2 with IC50 values of 2 nM and 52 nM for EZH2 and EZH1 respectively. It has potential antineoplastic activity.
Catalog No. 50-238-9922 Supplier Selleck Chemical LLC Supplier No. S83532MG
Lirametostat (CPI-1205) is an orally available selective inhibitor of the histone lysine methyltransferase EZH2 with IC50 values of 2 nM and 52 nM for EZH2 and EZH1 respectively. It has potential antineoplastic activity.