Learn More
Medchemexpress LLC HY-19672 5mg Medchemexpress, Lirimilast CAS:329306-27-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY196725MG
Encompass_Preferred
Medchemexpress, HY-19672 5mg Lirimilast CAS:329306-27-6 Lirimilast (BAY 19-8004) is a potent, selective and orally active phosphodiesterase-4 (PDE4) inhibitor with an IC50 value of 49 nM. Lirimilast can be used for the treatment of asthma or chronic obstructive pulmonary disease (COPD). Lirimilast has potently anti-inflammatory properties. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-197-0030