Learn More
Cayman Chemical Lisinopril, 76547-98-3, 500 mg
SDP
Supplier:  Cayman Chemical 16833500
Encompass_Preferred
A potent ACE inhibitor that blocks the formation of angiotensin I with an IC50 value of 1.2 nM (ovine) and displays ID50 values of 2.3 and 6.5 µg/kg in rat and canine, respectively
Catalog No. NC0981721