Learn More
Medchemexpress LLC Lisinopril | 76547-98-3 | 100.0% | 405.49 | 250 MG
SDP
Catalog No. 5000384396
Encompass_Preferred
Lisinopril (MK-521) is an angiotensin-converting enzyme inhibitor used in the treatment of hypertension, congestive heart failure, and heart attacks. It is a potent, competitive inhibitor of angiotensin-converting enzyme (ACE), the enzyme responsible for the conversion of angiotensin I to angiotensin II, which regulates blood pressure and is a key component of the renin-angiotensin-aldosterone system (RAAS).
  • Used to treat hypertension and symptomatic congestive heart failure.
  • Improves survival in certain individuals following myocardial infarction.
  • Prevents the progression of renal disease in hypertensive patients with diabetes mellitus and microalbuminuria or overt nephropathy.
Catalog No. 50-003-84396 Supplier Medchemexpress LLC Supplier No. 250MGHY18206
Lisinopril (MK-521) is an angiotensin-converting enzyme inhibitor used in the treatment of hypertension, congestive heart failure, and heart attacks. It is a potent, competitive inhibitor of angiotensin-converting enzyme (ACE), the enzyme responsible for the conversion of angiotensin I to angiotensin II, which regulates blood pressure and is a key component of the renin-angiotensin-aldosterone system (RAAS).
  • Used to treat hypertension and symptomatic congestive heart failure.
  • Improves survival in certain individuals following myocardial infarction.
  • Prevents the progression of renal disease in hypertensive patients with diabetes mellitus and microalbuminuria or overt nephropathy.