Learn More
AdipoGen Lisofylline
SDP
Catalog No. 502560716
Encompass_Preferred
Chemical. CAS 6493-06-7. Formula C13H20N4O3. MW 280.3. Synthetic. Synthetic methylxanthine metabolite of pentoxifylline. Potent anti-inflammatory agent in which only the − optical isomer is biologically active. Inhibits the generation of phosphatidic acid from cytokine-activated lysophosphatidic acyl transferase LPAAT, which has been shown to protect mice from endotoxic shock. Decreases lipid peroxidation in vitr oand in vivo. Suppresses the production of the proinflammatory cytokine IFN-gamma, inhibits IL-12-mediated STAT-4 activation, enhances glucose-stimulated beta-cell insulin secretion, reducing the onset of diabetes in a non-obese diabetic mouse model, and blocks autoimmune deterioration of pancreatic beta cells in non-obese diabetic mice. Compound can be used as analytical reference material.
Catalog No. 50-256-0716 Supplier AdipoGen Supplier No. CDXH0117M050
Chemical. CAS 6493-06-7. Formula C13H20N4O3. MW 280.3. Synthetic. Synthetic methylxanthine metabolite of pentoxifylline. Potent anti-inflammatory agent in which only the − optical isomer is biologically active. Inhibits the generation of phosphatidic acid from cytokine-activated lysophosphatidic acyl transferase LPAAT, which has been shown to protect mice from endotoxic shock. Decreases lipid peroxidation in vitr oand in vivo. Suppresses the production of the proinflammatory cytokine IFN-gamma, inhibits IL-12-mediated STAT-4 activation, enhances glucose-stimulated beta-cell insulin secretion, reducing the onset of diabetes in a non-obese diabetic mouse model, and blocks autoimmune deterioration of pancreatic beta cells in non-obese diabetic mice. Compound can be used as analytical reference material.