Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (+/-)-Lisofylline-d6 | 1185995-26-9 | ≥98.0% | 286.36 g/mol | C13H14D6N4O3 | 1MG
SDP

Catalog No. 5000235922
Encompass_Preferred

(±)-Lisofylline-d6 is the deuterium-labeled racemic form of lisofylline supplied as a neat stable-isotope reference material for research and analytical use. It is intended as an internal standard or tracer in pharmacokinetic, metabolic, and bioanalytical studies and is provided in small laboratory quantities.

  • Isotopically labeled with six deuterium atoms.
  • Racemic (±) mixture of lisofylline enantiomers.
  • Cas number 1185995-26-9.
  • Chemical formula C13H14D6N4O3.
  • Molecular weight 286.36 g/mol.
  • Purity ≥98.0% by HPLC; deuterium enrichment ≥99% atom D.
  • Supplied as a neat solid in a 1 mg pack.
  • Store at -20°C; shipped at room temperature.
  • For research use only; not for human or clinical use.

Catalog No. 50-002-35922 Supplier Medchemexpress LLC Supplier No. HY126042S1MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

(±)-Lisofylline-d6 is the deuterium-labeled racemic form of lisofylline supplied as a neat stable-isotope reference material for research and analytical use. It is intended as an internal standard or tracer in pharmacokinetic, metabolic, and bioanalytical studies and is provided in small laboratory quantities.

  • Isotopically labeled with six deuterium atoms.
  • Racemic (±) mixture of lisofylline enantiomers.
  • Cas number 1185995-26-9.
  • Chemical formula C13H14D6N4O3.
  • Molecular weight 286.36 g/mol.
  • Purity ≥98.0% by HPLC; deuterium enrichment ≥99% atom D.
  • Supplied as a neat solid in a 1 mg pack.
  • Store at -20°C; shipped at room temperature.
  • For research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.